APO455837016 Apollo Scientific Ltd

4-(Aminomethyl)-2-(difluoromethyl)pyridine, 4-BOC protected

  • MDL number MFCD23701108
  • Cas Number : 1428532-94-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym tert-Butyl {[2-(difluoromethyl)pyridin-4-yl]methyl}carbamate, 4-{[(tert-Butoxycarbonyl)amino]methyl}-2-(difluoromethyl)pyridine
Purity 95
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=52280

Safety Information

signalword Warning
pcodes P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=52280
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart