APO455837021 Apollo Scientific Ltd

4-Chloro-6-(trifluoromethyl)nicotinic acid

  • MDL number MFCD13188812
  • Cas Number : 1060810-66-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Chloro-6-(trifluoromethyl)pyridine-3-carboxylic acid, 5-Carboxy-4-chloro-2-(trifluoromethyl)pyridine
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=52916

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=52916
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart