APO455837025 Apollo Scientific Ltd

4-Hydroxy-5-iodo-2-(trifluoromethyl)pyridine

  • MDL number MFCD24369747
  • Cas Number : 1027818-89-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Iodo-2-(trifluoromethyl)pyridin-4-ol, 4-Hydroxy-5-iodo-alpha,alpha,alpha-trifluoro-2-picoline
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=52920

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=52920
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart