APO455837027 Apollo Scientific Ltd

5-Amino-4-chloro-2-(trifluoromethyl)pyridine

  • MDL number MFCD11848353
  • Cas Number : 1196153-86-2
  • Molecular Weight : 196.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-chloro-6-(trifluoromethyl)-3-pyridinylamine
Inchi Code 1S/C6H4ClF3N2/c7-3-1-5(6(8,9)10)12-2-4(3)11/h1-2H,11H2
Inchi Key XKRQUYRYIFNHNV-UHFFFAOYSA-N

Description

Synonym 4-Chloro-6-(trifluoromethyl)pyridin-3-amine, 5-Amino-4-chloro-alpha,alpha,alpha-trifluoro-2-picoline
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-amino-4-chloro-2-trifluoromethylpyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-amino-4-chloro-2-trifluoromethylpyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart