APO455837033 Apollo Scientific Ltd

5-Methyl-2-(trifluoromethyl)fur-3-oylacetonitrile

  • MDL number MFCD00277690
  • Cas Number : 175276-72-9
  • Molecular Weight : 217.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75-76
Synonym 3-(Cyanoacetyl)-5-methyl-2-(trifluoromethyl)furan
Purity 97
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-methyl-2-trifluoromethylfur-3-oylacetonitrile

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/5-methyl-2-trifluoromethylfur-3-oylacetonitrile
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart