APO455837034 Apollo Scientific Ltd

Methyl pentafluoropropionylacetate

  • MDL number MFCD00041000
  • Cas Number : 104857-88-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 124-126
Product Link http://www.apolloscientific.co.uk/display_item.php?id=3959

Safety Information

signalword Warning
pcodes P210;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=3959
pictograms GHS02
hstatements H226
Availability details for
Quantity
  • Availability
  • Add to Cart