APO455837035 Apollo Scientific Ltd

5-Amino-2-fluorophenylacetic acid

  • MDL number MFCD03094259
  • Cas Number : 518057-74-4
  • Molecular Weight : 169.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-137
Synonym 3-(Carboxymethyl)-4-fluoroaniline
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-amino-2-fluorophenylacetic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-amino-2-fluorophenylacetic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart