APO455837047 Apollo Scientific Ltd

4-Chloro-5-(trifluoromethyl)benzene-1,2-diamine

  • MDL number MFCD00119550
  • Cas Number : 157590-59-5
  • Molecular Weight : 210.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 61-63
Synonym 4-Chloro-5-(trifluoromethyl)phenylene-1,2-diamine
Purity 97
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-chloro-5-trifluoromethylbenzene-12-diamine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-chloro-5-trifluoromethylbenzene-12-diamine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart