APO455837048 Apollo Scientific Ltd

2-Amino-5-fluoro-3-nitrobenzotrifluoride

  • MDL number MFCD03094266
  • Cas Number : 344-29-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Fluoro-2-nitro-6-(trifluoromethyl)aniline, 2-Amino-5-fluoro-3-(trifluoromethyl)nitrobenzene
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-amino-5-fluoro-3-nitrobenzotrifluoride-97

Safety Information

signalword Warning
pcodes P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-5-fluoro-3-nitrobenzotrifluoride-97
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart