APO455837049 Apollo Scientific Ltd

2-Methyl-4,4,4-trifluorobutanol

  • MDL number MFCD00190639
  • Cas Number : 107103-95-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 134
Product Link http://www.apolloscientific.co.uk/display_item.php?id=3998

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=3998
pictograms GHS02;GHS07
hstatements H226;H302;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart