APO455837052 Apollo Scientific Ltd

4,5-Diamino-2-fluorobenzotrifluoride

  • MDL number MFCD04038250
  • Cas Number : 179062-06-7
  • Molecular Weight : 194.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 48-51
Synonym 4-Fluoro-5-(trifluoromethyl)benzene-1,2-diamine, 1,2-Diamino-4-fluoro-5-(trifluoromethyl)benzene
Purity 97
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/45-diamino-2-fluorobenzotrifluoride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/45-diamino-2-fluorobenzotrifluoride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart