APO455837055 Apollo Scientific Ltd

Methyl 2-(trifluoromethyl)benzoate

  • MDL number MFCD00236050
  • Cas Number : 344-96-7
  • Molecular Weight : 204.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 1.30
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 98-100
Product Link https://store.apolloscientific.co.uk/product/methyl-2-trifluoromethylbenzoate

Safety Information

signalword Warning
pcodes P280;P304+P340
MSDS https://store.apolloscientific.co.uk/product/methyl-2-trifluoromethylbenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart