APO455837058 Apollo Scientific Ltd

5-Bromo-3-fluorobenzene-1,2-diamine

  • MDL number MFCD03094270
  • Cas Number : 517920-69-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Bromo-1,2-diamino-3-fluorobenzene
Purity 97
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2421

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=2421
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart