APO455837062 Apollo Scientific Ltd

Methyl 2H-hexafluoro-2-methylpropanoate

  • MDL number MFCD00054663
  • Cas Number : 360-54-3
  • Molecular Weight : 210.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 60
Iupac Name methyl 3,3,3-trifluoro-2-(trifluoromethyl)propanoate
density 1.604
Inchi Code 1S/C5H4F6O2/c1-13-3(12)2(4(6,7)8)5(9,10)11/h2H,1H3
Inchi Key JGGBVFLRNNDHCM-UHFFFAOYSA-N

Description

Melting Point 60
Synonym Methyl 3,3,3-trifluoro-2-(trifluoromethyl)propanoate, Methyl 3,3,3-trifluoro-2-(trifluoromethyl)propionate
Purity 98%
Density 1.604
Flash Point 90-91
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-2h-hexafluoro-2-methylpropanoate

Safety Information

signalword Warning
pcodes P210;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-2h-hexafluoro-2-methylpropanoate
pictograms GHS02
hstatements H226
Availability details for
Quantity
  • Availability
  • Add to Cart