APO455837064 Apollo Scientific Ltd

Methyltrifluorosilane

  • MDL number MFCD00040332
  • Cas Number : 373-74-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -73
Purity 98
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point -30
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4107

Safety Information

signalword Warning
pcodes P210;P280;P304+P340
MSDS http://www.apolloscientific.co.uk/display_item.php?id=4107
pictograms GHS02;GHS07;GHS04
hstatements H226;H315;H319;H335;H280
Availability details for
Quantity
  • Availability
  • Add to Cart