APO455837068 Apollo Scientific Ltd

4-(2,2,2-Trifluoroethoxy)aniline

  • MDL number MFCD00077606
  • Cas Number : 57946-61-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63-66
Synonym 4-Amino-beta,beta,beta-trifluorophenetole
Purity 98
Flash Point >110
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5098

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=5098
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart