APO455837072 Apollo Scientific Ltd

Methyl 3-fluoro-4-iodobenzoate

  • MDL number MFCD09800728
  • Cas Number : 1027513-46-7
  • Molecular Weight : 280.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-3-fluoro-4-iodobenzoate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-3-fluoro-4-iodobenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart