APO455837078 Apollo Scientific Ltd

2-Fluoro-6-methoxybenzenesulphonyl chloride

  • MDL number MFCD12922637
  • Cas Number : 1176126-31-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(Chlorosulphonyl)-3-fluoroanisole
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=62703

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=62703
pictograms GHS05
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart