APO455837080 Apollo Scientific Ltd

3-Fluoro-5-(trifluoromethoxy)benzaldehyde

  • MDL number MFCD18783220
  • Cas Number : 1352999-98-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=62867

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=62867
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart