APO455837082 Apollo Scientific Ltd

5-Fluoro-2-formylbenzeneboronic acid

  • MDL number MFCD09953731
  • Cas Number : 1256355-30-2
  • Molecular Weight : 167.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 137-140
Synonym 2-Borono-4-fluorobenzaldehyde
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-fluoro-2-formylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-fluoro-2-formylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart