APO455837086 Apollo Scientific Ltd

Nickel(II) fluoride, anhydrous

  • MDL number MFCD00016263
  • Cas Number : 10028-18-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 1380
Purity 99
Density 4.72
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4118

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=4118
pictograms GHS07;GHS08;GHS09
hstatements H335;H360D;H410
Availability details for
Quantity
  • Availability
  • Add to Cart