APO455837087 Apollo Scientific Ltd

3,3,3-Trifluoro-DL-alanine methyl ester hydrochloride

  • MDL number MFCD08532471
  • Cas Number : 134297-36-2
  • Molecular Weight : 193.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 168-170
Synonym Methyl 2-amino-3,3,3-trifluoropropanoate hydrochloride, Methyl 3,3,3-trifluoro-DL-alaninate hydrochloride
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/333-trifluoro-dl-alanine-methyl-ester-hydrochloride

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/333-trifluoro-dl-alanine-methyl-ester-hydrochloride
pictograms GHS06
hstatements H301+H311+H331
Availability details for
Quantity
  • Availability
  • Add to Cart