APO455837092 Apollo Scientific Ltd

5,7-Bis(trifluoromethyl)-4-hydroxyquinoline-3-carboxylic acid

  • MDL number MFCD00269347
  • Cas Number : 240408-95-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=363

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=363
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart