APO455837095 Apollo Scientific Ltd

5-Fluoropyridine-2-carboxylic acid

  • MDL number MFCD04114196
  • Cas Number : 107504-08-5
  • Molecular Weight : 141.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164-166
Synonym 5-Fluoropicolinic acid, 2-Carboxy-5-fluoropyridine
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-fluoropyridine-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-fluoropyridine-2-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart