APO455837100 Apollo Scientific Ltd

2-Hydroxy-5-(trifluoromethoxy)benzoic acid

  • MDL number MFCD01091003
  • Cas Number : 129644-57-1
  • Molecular Weight : 222.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 130-132
Iupac Name 2-hydroxy-5-(trifluoromethoxy)benzoic acid
Inchi Code 1S/C8H5F3O4/c9-8(10,11)15-4-1-2-6(12)5(3-4)7(13)14/h1-3,12H,(H,13,14)
Inchi Key HNYMLXYADOZCNY-UHFFFAOYSA-N

Description

Synonym 5-(Trifluoromethoxy)salicylic acid, 2-Carboxy-4-(trifluoromethoxy)phenol
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 130-132
Product Link https://store.apolloscientific.co.uk/product/2-hydroxy-5-trifluoromethoxybenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-hydroxy-5-trifluoromethoxybenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart