APO455837101 Apollo Scientific Ltd

2-Amino-3-(trifluoromethoxy)benzoic acid

  • MDL number MFCD08669644
  • Cas Number : 561304-41-4
  • Molecular Weight : 221.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99-101
Synonym 3-(Trifluoromethoxy)anthranilic acid, 2-Carboxy-6-(trifluoromethoxy)aniline
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-amino-3-trifluoromethoxybenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-3-trifluoromethoxybenzoic-acid
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart