APO455837102 Apollo Scientific Ltd

Ethyl 2-hydroxy-4-methyl-2-(trifluoromethyl)pent-4-enoate

  • MDL number MFCD08532486
  • Cas Number : 217195-91-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(Ethoxycarbonyl)-4-methyl-1,1,1-trifluoropent-4-en-2-ol
Purity 97
Density 1.194
Flash Point >110
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 46/2.3mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=10972

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=10972
pictograms GHS07
hstatements H302;H315;H319;H332
Availability details for
Quantity
  • Availability
  • Add to Cart