APO455837103 Apollo Scientific Ltd

Perfluoropentyl iodide

  • MDL number MFCD08461629
  • Cas Number : 638-79-9
  • Molecular Weight : 395.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -50
Synonym Perfluoro-1-iodopentane, 1-Iodo-1,1,2,2,3,3,4,4,5,5,5-undecafluoropentane, 1-Iodoperfluoropentane
Purity 98
Density 2.06
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 94.4
Product Link https://store.apolloscientific.co.uk/product/perfluoropentyl-iodide

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/perfluoropentyl-iodide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart