APO455837104 Apollo Scientific Ltd

2-Fluoro-3-nitrobenzotrifluoride

  • MDL number MFCD08532487
  • Cas Number : 61324-97-8
  • Molecular Weight : 209.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-fluoro-1-nitro-3-(trifluoromethyl)benzene
Inchi Code 1S/C7H3F4NO2/c8-6-4(7(9,10)11)2-1-3-5(6)12(13)14/h1-3H
Inchi Key PGKWPKDZNHZBQK-UHFFFAOYSA-N

Description

Synonym 2-Fluoro-3-(trifluoromethyl)nitrobenzene, 2-Fluoro-1-nitro-3-(trifluoromethyl)benzene
Purity 98%
Storage temp. Ambient Temperature
Physical Form Liquid-Clear
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-3-nitrobenzotrifluoride

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-3-nitrobenzotrifluoride
pictograms GHS06
hstatements H301+H311+H331
Availability details for
Quantity
  • Availability
  • Add to Cart