APO455837105 Apollo Scientific Ltd

2-Hydroxy-3-nitrobenzotrifluoride

  • MDL number MFCD05155649
  • Cas Number : 1548-62-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73-75
Synonym 2-Hydroxy-3-(trifluoromethyl)nitrobenzene, 2-Nitro-6-(trifluoromethyl)phenol
Purity 99
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-hydroxy-3-nitrobenzotrifluoride

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-hydroxy-3-nitrobenzotrifluoride
pictograms GHS06
hstatements H301+H311;H315;H319;H330;H335
Availability details for
Quantity
  • Availability
  • Add to Cart