APO455837106 Apollo Scientific Ltd

2-Methoxy-3-nitrobenzotrifluoride

  • MDL number MFCD08532488
  • Cas Number : 627531-40-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Methoxy-3-(trifluoromethyl)nitrobenzene, 2-Nitro-6-(trifluoromethyl)anisole
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=10978

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=10978
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart