APO455837119 Apollo Scientific Ltd

2'-Nitro-4'-(trifluoromethoxy)acetanilide

  • MDL number MFCD00052324
  • Cas Number : 787-57-5
  • Molecular Weight : 264.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94-96
Synonym N-[2-Nitro-4-(trifluoromethoxy)phenyl]acetamide, 2-(Acetylamino)-5-(trifluoromethoxy)nitrobenzene
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-nitro-4-trifluoromethoxyacetanilide

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-nitro-4-trifluoromethoxyacetanilide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart