APO455837123 Apollo Scientific Ltd

3-Nitro-5-(trifluoromethyl)benzoic acid

  • MDL number MFCD00024509
  • Cas Number : 328-80-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 127-132
Synonym 3-Carboxy-5-nitrobenzotrifluoride, 3-Carboxy-5-(trifluoromethyl)nitrobenzene
Purity 97
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-nitro-5-trifluoromethylbenzoic-acid-97

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-nitro-5-trifluoromethylbenzoic-acid-97
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart