APO455837129 Apollo Scientific Ltd

3-Chloro-2-fluorophenylhydrazine hydrochloride

  • MDL number MFCD03094280
  • Cas Number : 517920-75-1
  • Molecular Weight : 197.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Chloro-2-fluoro-3-hydrazinobenzene hydrochloride
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-chloro-2-fluorophenylhydrazine-hydrochloride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloro-2-fluorophenylhydrazine-hydrochloride
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart