APO455837134 Apollo Scientific Ltd

3-Bromo-2-chloro-6-fluorobenzoic acid

  • MDL number MFCD11856020
  • Cas Number : 1114809-13-0
  • Molecular Weight : 253.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134-136
Storage temp. Store at 2-8
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-2-chloro-6-fluorobenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-2-chloro-6-fluorobenzoic-acid
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart