APO455837136 Apollo Scientific Ltd

3'-Bromo-2'-chloro-6'-fluoroacetophenone

  • MDL number MFCD28140553
  • Cas Number : 1704367-13-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(3-Bromo-2-chloro-6-fluorophenyl)ethanone
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=66979

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=66979
pictograms GHS07
hstatements H302;H312;H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart