APO455837140 Apollo Scientific Ltd

5-Fluoro-2-(2,2,2-trifluoroethoxy)nitrobenzene

  • MDL number MFCD28155023
  • Cas Number : 334929-98-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Fluoro-2-nitro-1-(2,2,2-trifluoroethoxy)benzene, 4-Fluoro-2-nitrophenyl 2,2,2-trifluoroethyl ether, 2-Nitro-beta,beta,beta,4-tetrafluorophenetole
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=67026

Safety Information

signalword Warning
pcodes P260;P280;P312
MSDS http://www.apolloscientific.co.uk/display_item.php?id=67026
pictograms GHS07
hstatements H302;H312;H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart