APO455837141 Apollo Scientific Ltd

6-Fluoro-4-(trifluoromethyl)-1H-benzimidazole

  • MDL number MFCD28155024
  • Cas Number : 1360936-21-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=67027

Safety Information

signalword Warning
pcodes P260;P280;P312
MSDS http://www.apolloscientific.co.uk/display_item.php?id=67027
pictograms GHS07
hstatements H302;H312;H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart