APO455837145 Apollo Scientific Ltd

1-Fluorononane

  • MDL number MFCD00000455
  • Cas Number : 463-18-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym n-Nonyl fluoride
Purity 98
Density 0.81
Flash Point 48
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 166-169/760mmHg
Product Link http://www.apolloscientific.co.uk/display_item.php?id=1975

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=1975
pictograms GHS02;GHS06
hstatements H226;H300;H311;H331;H335
Availability details for
Quantity
  • Availability
  • Add to Cart