APO455837149 Apollo Scientific Ltd

1H,1H-Perfluorobutane

  • MDL number MFCD00236695
  • Cas Number : 662-35-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1H,1H-Octafluorobutane
Purity 97
Storage temp. Ambient Temperature
Physical Form Gas
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4203

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=4203
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart