APO455837152 Apollo Scientific Ltd

4-(Trifluoromethyl)phenylhydrazine hydrochloride

  • MDL number MFCD00204233
  • Cas Number : 2923-56-0
  • Molecular Weight : 212.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-trifluoromethylphenylhydrazine-hydrochloride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-trifluoromethylphenylhydrazine-hydrochloride
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart