APO455837158 Apollo Scientific Ltd

5H-Octafluoropentanoyl chloride

  • MDL number MFCD00153221
  • Cas Number : 376-71-6
  • Molecular Weight : 264.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5H-Octafluorovaleroyl chloride, 2,2,3,3,4,4,5,5-Octafluoropentanoyl chloride
Purity 97
Density 1.67
Storage temp. Store at 2-8
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 85-88
Product Link https://store.apolloscientific.co.uk/product/5h-octafluoropentanoyl-chloride

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5h-octafluoropentanoyl-chloride
pictograms GHS05;GHS07
hstatements H314;H335
Availability details for
Quantity
  • Availability
  • Add to Cart