APO455837162 Apollo Scientific Ltd

2,4-Difluoro-DL-phenylglycine

  • MDL number MFCD02662503
  • Cas Number : 240409-02-3
  • Molecular Weight : 187.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Amino(2,4-difluorophenyl)acetic acid, 2-Amino-2-(2,4-difluorophenyl)ethanoic acid
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/24-difluoro-dl-phenylglycine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/24-difluoro-dl-phenylglycine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart