APO455837168 Apollo Scientific Ltd

3-(Trifluoromethyl)phenacyl bromide

  • MDL number MFCD03094283
  • Cas Number : 2003-10-3
  • Molecular Weight : 267.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 20-23
Synonym 2-Bromo-3'-(trifluoromethyl)acetophenone, 2-Bromo-1-[3-(trifluoromethyl)phenyl]ethan-1-one, 3-(Bromoacetyl)benzotrifluoride
Storage temp. Store at 2-8
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 64-72/0.07mm
Product Link https://store.apolloscientific.co.uk/product/3-trifluoromethylphenacyl-bromide

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-trifluoromethylphenacyl-bromide
pictograms GHS05;GHS07
hstatements H314;H335
Availability details for
Quantity
  • Availability
  • Add to Cart