APO455837173 Apollo Scientific Ltd

2,4-Dihydro-4-methyl-5-(trifluoromethyl)-3H-1,2,4-triazole-3-thione

  • MDL number MFCD00003095
  • Cas Number : 30682-81-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 116-118
Synonym 2,4-Dihydro-4-methyl-3-thioxo-5-(trifluoromethyl)-3H-1,2,4-triazole
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4097

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=4097
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart