APO455837191 Apollo Scientific Ltd

2,2-Bis[4-(4-nitrophenoxy)phenyl]hexafluoropropane

  • MDL number MFCD00317556
  • Cas Number : 69563-87-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/22-bis4-4-nitrophenoxyphenylhexafluoropropane

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/22-bis4-4-nitrophenoxyphenylhexafluoropropane
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart