APO455837193 Apollo Scientific Ltd

Pentafluoroanisole

  • MDL number MFCD00000289
  • Cas Number : 389-40-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -38
Purity 98
Density 1.398
Flash Point 32
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 136-138
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4252

Safety Information

signalword Warning
pcodes P210;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=4252
pictograms GHS02
hstatements H226
Availability details for
Quantity
  • Availability
  • Add to Cart