APO455837195 Apollo Scientific Ltd

2-(Trifluoromethyl)-DL-phenylglycine

  • MDL number MFCD02662403
  • Cas Number : 240490-00-0
  • Molecular Weight : 219.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Amino[2-(trifluoromethyl)phenyl]acetic acid, 2-[Amino(carboxy)methyl]benzotrifluoride
Purity 97
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-trifluoromethyl-dl-phenylglycine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-trifluoromethyl-dl-phenylglycine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart