APO455837201 Apollo Scientific Ltd

2-Chloro-6-(trifluoromethyl)nicotinonitrile

  • MDL number MFCD01862660
  • Cas Number : 386704-06-9
  • Molecular Weight : 206.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 36-38
Synonym 2-Chloro-6-(trifluoromethyl)pyridine-3-carbonitrile
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 74-76/0.8mm
Product Link https://store.apolloscientific.co.uk/product/2-chloro-6-trifluoromethylnicotinonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-6-trifluoromethylnicotinonitrile
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart