APO455837205 Apollo Scientific Ltd

5-[3-(Trifluoromethyl)phenyl]-1H-tetrazole

  • MDL number MFCD00068098
  • Cas Number : 92712-48-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156-158
Synonym 3-(1H-Tetrazol-5-yl)benzotrifluoride
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5442

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=5442
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart